missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Liquid Scintillation Vials, PET
Provides the clarity of glass with the safety of plastic.
Brand: DWK Life Sciences 986740
This item is not returnable.
View return policy
Description
Provides the clarity of glass with the safety of plastic.
- Vials offer low permeability to solvents and minimal background counts
- Clarity of glass with the safety of plastic
- Vials can be safely incinerated
- No harmful gas is generated
- Packaged in 5 utility trays with each partition tray holding 100 vials
- Utility trays serve as a way to store your samples
Specifications
Foamed Polyethylene | |
27 mm | |
20 mL | |
Polypropylene | |
22-400 |
Clear | |
PET | |
1000 | |
61 mm |