missing translation for 'onlineSavingsMsg'
Learn More

Lisuride maleate, Tocris Bioscience™

Product Code. 18712594
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

DA receptor agonist; antiparkinson's agent.

Dopamine receptor agonist and anti-Parkinson's agent. Displays high affinity for D2, D3 and D4 receptors along with 5-HT1A. Exhibits some 5-HT2B receptor antagonist properties. Decreases prolactin release; reduces inflammatory mediators such as TNF-α and IL6. Exhibits anticonvulsive effects. Acts similar to bromocriptine (Cat. No. 0427).

Chemical Identifiers

CAS 19875-60-6
Molecular Formula C24H30N4O5
Molecular Weight (g/mol) 454.527
InChI Key CVQFAMQDTWVJSV-BAXNFHPCSA-N
Synonym lisuride maleate, 1,1-diethyl-3-8alpha-6-methyl-9,10-didehydroergolin-8-yl urea 2z-but-2-enedioate
PubChem CID 25137867
ChEBI CHEBI:31776
IUPAC Name 3-[(6aR,9S)-7-methyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-yl]-1,1-diethylurea;(Z)-but-2-enedioate;hydron
SMILES [H+].[H+].CCN(CC)C(=O)NC1CN(C2CC3=CNC4=CC=CC(=C34)C2=C1)C.C(=CC(=O)[O-])C(=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 446.55
Percent Purity >99%
Chemical Name or Material Lisuride maleate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'