missing translation for 'onlineSavingsMsg'
Learn More

Lithium 2-(morpholinomethyl)benzoate , Tech., Thermo Scientific™

Product Code. 10749605
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 904696-60-2
CAS Min % 80.0
CAS Max % 100.0
Molecular Formula C12H14LiNO3
MDL Number MFCD08690277
Quantity 1 g
Synonym lithium 2-morpholinomethyl benzoate, lithium 2-morpholin-4-ylmethyl benzoate, lithium 1+ ion 2-morpholin-4-ylmethyl benzoate, lithium 1+ 2-morpholin-4-ylmethyl benzoate, lithotab 2-morpholin-4-ylmethyl benzoate, lithium 2-morpholin-4-yl methyl benzoate, lithium 2-morpholin-4-ylmethyl benzoate, tech, 2-morpholin-4-ylmethyl benzoic acid, lithium salt, lithium 1+ ion 2-morpholin-4-yl methyl benzoate, benzoicacid,2-4-morpholinylmethyl-,lithium salt 1:1
InChI Key PKLIDBHLNQBFTO-UHFFFAOYSA-M
SMILES [Li+].C1COCCN1CC2=CC=CC=C2C(=O)[O-]
IUPAC Name lithium;2-(morpholin-4-ylmethyl)benzoate
Molecular Weight (g/mol) 227.188
PubChem CID 23713561
Formula Weight 227.19
Percent Purity Tech.
Grade Technical
Chemical Name or Material Lithium 2-(morpholinomethyl)benzoate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'