missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals

Product Code. 15157442
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H3LiO2·2H2O
Molecular Weight (g/mol) 102.02
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 56.0°C to 58.0°C
Quantity 250 g
Assay Percent Range 98%
Linear Formula CH3CO2Li·2H2O
Merck Index 15,5577
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: very soluble in hot water, 215g/L ethanol
Formula Weight 102.02
Percent Purity 98%
Grade Analytical
Packaging Plastic Bottle
Chemical Name or Material Lithium Acetate Dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium acetate dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'