missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetylide ethylenediamine complex, 85%

Product Code. 10206100 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 10206100

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6867-30-7 | C4H9LiN2 | 92.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6867-30-7
Molecular Formula C4H9LiN2
Molecular Weight (g/mol) 92.07
MDL Number MFCD00013183
InChI Key QJQWXTYPTBEPGS-UHFFFAOYSA-N
Synonym lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
PubChem CID 2724010
IUPAC Name lithium;ethane-1,2-diamine;ethyne
SMILES [Li]C#C.NCCN

Specifications

Melting Point 76.0°C
Color Yellow
Boiling Point 110.6°C
Assay Percent Range 80% min. (Gasvolumetric)
Linear Formula HC≡CLi·H2NCH2CH2NH2
Quantity 50 g
Fieser 01,574; 05,382; 07,195; 11,288; 01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59
Solubility Information Solubility in water: vigorous reaction. Other solubilities: soluble in ethylene diamine,n-propylamine and,pyridine
Formula Weight 92.07
Percent Purity 85%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Lithium acetylide ethylenediamine complex, 85%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium acetylide ethylenediamine complex
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Ethanediamine, compound with lithiumacetylide (Li(C2H))(1:1)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.