missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium aluminum hydride, 95%, powder, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
100 g
500 g
5 kg
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Spezifikation
Spezifikation
Melting Point | 125.0°C |
Physical Form | Powder |
Quantity | 5 kg |
Assay Percent Range | 95% |
Linear Formula | LiAlH4 |
Fieser | 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162 |
Merck Index | 15, 344 |
Solubility Information | Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd |
InChI Key | BJKLPLABXHXMIM-UHFFFAOYSA-N |
SMILES | [H-].[H-].[H-].[H-].[Li+].[Al+3] |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'