missing translation for 'onlineSavingsMsg'
Learn More

Lithium bis(trimethylsilyl)amide, 0.9M solution in methylcyclohexane, AcroSeal™, Thermo Scientific™

Product Code. 10472293
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4039-32-1, 108-87-2
MDL Number MFCD00008261
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
PubChem CID 2733832
IUPAC Name lithium;bis(trimethylsilyl)azanide
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C

Specifications

Color Colorless to Yellow
Density 0.7700g/mL
Flash Point −3°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H18LiNSi2
Quantity 800 mL
Specific Gravity 0.77
Molecular Weight (g/mol) 167.327
Formula Weight 167.33
Physical Form Liquid
Chemical Name or Material Lithium bis(trimethylsilyl)amide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium bis(trimethylsilyl)amide, 0.9M solution in methylcyclohexane, AcroSeal™, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.