Learn More
Lithium bromide, 4M solution in THF, AcroSeal™, Thermo Scientific Chemicals
£167.00 - £632.00
Specifications
Chemical Name or Material | Lithium bromide |
---|---|
CAS | 7550-35-8, 109-99-9 |
Molecular Formula | BrLi |
Linear Formula | LiBr |
MDL Number | MFCD00011077 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10085003
|
Thermo Scientific Acros
390971000 |
100 mL | AcroSeal™ Glass bottle |
£167.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137043
|
Thermo Scientific Acros
390978000 |
800 mL | AcroSeal™ Glass Bottle |
£632.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium bromide | |
BrLi | |
MFCD00011077 | |
15, 5582 | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.84 | |
Clear colorless to yellowish solution | |
1.1800g/mL |
7550-35-8, 109-99-9 | |
LiBr | |
−22°C | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
[Li+].[Br-] | |
86.84 | |
CHEBI:63042 | |
4M exact strength on the certificate of analysis | |
1.18 |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
May cause an a
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a POISON CENTER or doctor/physician if you
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-439-8
- Lithium bromide, 4M solution in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only