Learn More
Lithium bromide, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
£26.30 - £1330.00
Chemical Identifiers
CAS | 7550-35-8 |
---|---|
Molecular Formula | BrLi |
Molecular Weight (g/mol) | 86.84 |
MDL Number | MFCD00011077 |
InChI Key | AMXOYNBUYSYVKV-UHFFFAOYSA-M |
Synonym | lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide |
PubChem CID | 82050 |
ChEBI | CHEBI:63042 |
IUPAC Name | lithium;bromide |
SMILES | [Li+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10741091
|
Thermo Scientific Acros
199871000 |
100 g |
£26.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10092470
|
Thermo Scientific Acros
199875000 |
500 g |
£93.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10164813
|
Thermo Scientific Acros
199870025 |
2.5 kg |
£396.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15234674
|
Thermo Scientific Acros
199870100 |
10 kg |
£1330.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7550-35-8 | |
86.84 | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
82050 | |
lithium;bromide |
BrLi | |
MFCD00011077 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
CHEBI:63042 | |
[Li+].[Br-] |
Specifications
1265.0°C | |
7550-35-8 | |
6 to 12 (4% soln. at 20°C) | |
99% min. (Argentometry) | |
LiBr | |
01,604; 02,245; 04,297 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.84 | |
Analytical | |
Plastic bottle | |
3.4600g/mL |
550.0°C | |
Gray to Black | |
Powder | |
BrLi | |
MFCD00011077 | |
15,5582 | |
Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol | |
[Li+].[Br-] | |
86.84 | |
CHEBI:63042 | |
≥99% | |
0.5% max. (400°C) | |
3.46 | |
Lithium bromide, 99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-439-8
missing translation for 'rtecsNumber' : OJ5755000
missing translation for 'tsca' : TSCA
- Lithium bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only