Learn More
Lithium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals
£51.66 - £447.23
Chemical Identifiers
CAS | 7550-35-8 |
---|---|
Molecular Formula | BrLi |
Molecular Weight (g/mol) | 86.84 |
InChI Key | AMXOYNBUYSYVKV-UHFFFAOYSA-M |
Synonym | lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide |
PubChem CID | 82050 |
ChEBI | CHEBI:63042 |
IUPAC Name | lithium;bromide |
SMILES | [Li+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15184319
|
Thermo Scientific Acros
453982500 |
250 g | Plastic Bottle |
£51.66
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15259754
|
Thermo Scientific Acros
453980010 |
1 kg | Plastic Bottle |
£168.84
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15299315
|
Thermo Scientific Acros
453980025 |
2.5 kg | Plastic Bottle |
£447.23
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
7550-35-8 | |
86.84 | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
CHEBI:63042 | |
[Li+].[Br-] |
BrLi | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
82050 | |
lithium;bromide |
Specifications
1265.0°C | |
7550-35-8 | |
98.5% min. (Argentometry) | |
LiBr | |
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide | |
AMXOYNBUYSYVKV-UHFFFAOYSA-M | |
lithium;bromide | |
82050 | |
86.84 | |
Pure | |
3.4600g/mL |
550.0°C | |
6 to 12 (4 % aq. soln.) | |
BrLi | |
15,5582 | |
Solubility in water: 1.49 kg/l (20°C). Other solubilities: slightly soluble in alcohol, ether, methanol,, acetone and glycol, slightly soluble in pyridine, soluble in amyl alcohol | |
[Li+].[Br-] | |
86.84 | |
CHEBI:63042 | |
99% | |
3.46 | |
Lithium bromide |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protect
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-439-8
- Lithium bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only