missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, SPEX CertiPrep™

Product Code. 15206086
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy
ultra pure, 1kg

Specifications

Quantity 1 kg
compliance-icons
Product Identifier
  • Fusion Flux - Ultra Pure Grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : lithium carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'