Learn More
Lithium carbonate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
£30.25 - £156.78
Chemical Identifiers
CAS | 554-13-2 |
---|---|
Molecular Formula | CLi2O3 |
Molecular Weight (g/mol) | 73.89 |
MDL Number | MFCD00011084 |
InChI Key | XGZVUEUWXADBQD-UHFFFAOYSA-L |
Synonym | lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine |
PubChem CID | 11125 |
ChEBI | CHEBI:6504 |
SMILES | [Li+].[Li+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092500
|
Thermo Scientific Acros
193360100 |
10 g | Glass bottle |
£30.25
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239833
|
Thermo Scientific Acros
193361000 |
100 g | Glass bottle |
£156.78
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
554-13-2 | |
73.89 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
11125 | |
[Li+].[Li+].[O-]C([O-])=O |
CLi2O3 | |
MFCD00011084 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
CHEBI:6504 |
Specifications
720.0°C | |
White | |
99.999% | |
Li2CO3 | |
01,606; 03,183; 04,192; 05,396 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
dilithium(1+) carbonate | |
11125 | |
73.88 | |
Trace Metal Basis | |
Lithium carbonate, Extra Pure |
554-13-2 | |
Powder | |
CLi2O3 | |
MFCD00011084 | |
15, 5583 | |
Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol | |
[Li+].[Li+].[O-]C([O-])=O | |
73.89 | |
CHEBI:6504 | |
99.999% | |
2.11g/mL |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-062-5
- Lithium carbonate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Lithium carbonate
RUO – Research Use Only