missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Carbonate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
100 kg
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Quantity | 1 kg |
Linear Formula | Li2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 3999191 |
Formula Weight | 73.89g/mol |
Percent Purity | ≥99% |
Grade | purum |
Chemical Name or Material | Lithium Carbonate |
Safety and Handling
Product Identifier
- Lithium carbonate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Lithium carbonate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'