missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, purum p.a, ≥98.0% (T), Honeywell™

Product Code. 15646050
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1300°C
Melting Point 720°C
Color White
pH 9 to 11 (at 1g/L, 20°C)
Physical Form Crystalline
Quantity 250 g
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
Formula Weight 73.89g/mol
Percent Purity ≥98.0% (T)
Odor Odorless
Packaging Poly bottle
Density 2.11g/cm3
Chemical Name or Material Lithium carbonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'