missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals

Product Code. 15244674
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Lithium Chloride, 99%, ClLi, CAS Number-7447-41-8, chlorure de lithium, chlorku litu polish, chlorku litu, lithium chloride licl, lithium chloride, licl, luthium chloride, lithiumchloride, lithiumchlorid, chlorolithium, 500g, carbonate (as Li2CO3): < 0.03% (typically), 98.5% min. (Argentometry)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382.0°C
Melting Point 605.0°C
Color White
Physical Form Crystalline Granules
Quantity 10 kg
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15,5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
Formula Weight 42.38
Percent Purity 99%
Grade Analytical
Loss on Drying 0.5% max. (400°C, 4 hrs)
Packaging Plastic drum
Specific Gravity 2.06
Density 2.0600g/mL
Chemical Name or Material Lithium chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.