missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride Anhydrous, Certified AR for Analysis, Fisher Chemical

Product Code. 10578430 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10578430

Brand: Fisher Chemical L/2201/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

250g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1382°C
Melting Point 605°C
CAS 7447-41-8
Color White
pH 7 to 8
Physical Form Solid
Quantity 250 g
Molecular Formula ClLi
MDL Number MFCD00011078
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES [Li+].[Cl-]
IUPAC Name lithium(1+) chloride
Molecular Weight (g/mol) 42.39
PubChem CID 433294
ChEBI CHEBI:48607
Formula Weight 42.38g/mol
Packaging Plastic powder jar
Vapor Pressure 1.33 hPa 547
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.