missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium chloride, Honeywell™
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Quantity | 500 g |
Linear Formula | LiCl |
UN Number | NONH for all modes of transport |
IUPAC Name | lithium;chloride |
Formula Weight | 42.39g/mol |
Percent Purity | ≥98% (AT) |
Chemical Name or Material | Lithium chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction