Learn More
Lithium chloride, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals
£53.80 - £496.00
Chemical Identifiers
CAS | 7447-41-8 |
---|---|
Molecular Formula | ClLi |
Molecular Weight (g/mol) | 42.39 |
MDL Number | MFCD00011078 |
InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
Synonym | lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride |
PubChem CID | 433294 |
ChEBI | CHEBI:48607 |
SMILES | [Li+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420011
|
Thermo Scientific Alfa Aesar
044762.06 |
5 g |
£53.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430011
|
Thermo Scientific Alfa Aesar
044762.14 |
25 g |
£184.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440011
|
Thermo Scientific Alfa Aesar
044762.22 |
100 g |
£496.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium chloride is useful for the production of lithium metal and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7447-41-8 | |
42.39 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
433294 | |
[Li+].[Cl-] |
ClLi | |
MFCD00011078 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
CHEBI:48607 |
Specifications
1325°C to 1360°C | |
7447-41-8 | |
Powder | |
ClLi | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
lithium(1+) chloride | |
433294 | |
42.39 | |
Hygroscopic | |
2.068 g/cm3 at 20°C |
605°C | |
White to Gray | |
99.995% (Metal basis) | |
MFCD00011078 | |
Soluble in water methanol,ethanol,pyridine and formic acid. Insoluble in dichloromethane. | |
[Li+].[Cl-] | |
42.39 | |
CHEBI:48607 | |
Odorless | |
White to gray crystalline powder | |
Lithium chloride, ultra dry |
Safety and Handling
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302-H315-H319
missing translation for 'einecsNumber' : 231-212-3
missing translation for 'rtecsNumber' : OJ5950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only