Learn More
Lithium cobalt(III) oxide, 97%, Thermo Scientific Chemicals
Description
Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Color | Blue |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 97% |
Solubility Information | Insoluble in water. |
Formula Weight | 97.88 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Dark blue powder |
Chemical Name or Material | Lithium cobalt(III) oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U29K021 | Certificate of Analysis | 30/09/2024 | 014049.18,014049.30,014049.A1 |
X18H024 | Certificate of Analysis | 12/04/2022 | 014049.18,014049.30,014049.A1 |
Q18D018 | Certificate of Analysis | 24/11/2021 | 014049.18,014049.30,014049.A1 |
A04Q01 | Certificate of Analysis | 24/11/2021 | 014049.18,014049.30,014049.A1 |
C31U029 | Certificate of Analysis | 24/11/2021 | 014049.18,014049.30,014049.A1 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.