missing translation for 'onlineSavingsMsg'
Learn More

Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane, AcroSeal™, Thermo Scientific™

Product Code. 11975911
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-84-6, 110-82-7
Molecular Formula C10H22LiNO
Molecular Weight (g/mol) 179.23
InChI Key YTJXGDYAEOTOCG-UHFFFAOYSA-N
Synonym lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran
PubChem CID 10997623
IUPAC Name lithium;di(propan-2-yl)azanide;oxolane
SMILES [Li+].CC(C)[N-]C(C)C.C1CCOC1

Specifications

Color Brown to Orange-Yellow
Physical Form Liquid
Quantity 100 mL
Flash Point −18°C
Formula Weight 179.23
Concentration or Composition (by Analyte or Components) 1.43 to 1.65M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.816
Density 0.8160g/mL
Chemical Name or Material Lithium diisopropylamide mono(tetrahydrofuran)
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'