missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 65.0°C to 67.0°C |
CAS | 65567-06-8, 109-99-9 |
Color | Orange to Red |
Physical Form | Liquid |
Quantity | 100 mL |
Molecular Formula | C12H10LiP |
MDL Number | MFCD00046054 |
Flash Point | −20°C |
Synonym | lithiodiphenylphosphine, lithium diphenylphosphide solution, diphenylphosphine lithium salt, lithium diphenylphosphanide, lipph2, lithiodiphenyl phosphide, lithium diphenylphosphine, diphenylphosphino lithium, diphenyl lithium phosphide |
InChI Key | WKUYEGHEUWHKIU-UHFFFAOYSA-N |
Show More |
Safety and Handling




Product Identifier
- Lithium diphenylphosphide, 0.5M solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium diphenylphosphide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction