missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lithium dodecyl sulfate, +99%

Product Code. 10186890
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym Dodecyl lithium sulfate, Lithium lauryl sulfate
PubChem CID 2735071
IUPAC Name lithium(1+) dodecyl sulfate
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Absorbance (5% aq. soln.) (1 cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm
Color White
Physical Form Shiny Crystalline Flakes or Powder
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99+%
Linear Formula CH3(CH2)11OSO3Li
Beilstein 01, IV, 1847
Solubility Information Solubility in water: soluble.
Formula Weight 272.33
Percent Purity ≥99%
Chemical Name or Material Lithium dodecyl sulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium dodecyl sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'