missing translation for 'onlineSavingsMsg'
Learn More

Lithium ethoxide, 1M solution in ethanol, Thermo Scientific Chemicals

Product Code. 10536843
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2388-07-0, 64-17-5
Molecular Formula C2H5LiO
Molecular Weight (g/mol) 52.00
MDL Number MFCD00050493
InChI Key AZVCGYPLLBEUNV-UHFFFAOYSA-N
Synonym lithium ethoxide, lithium ethylate, lithotab ethoxide, lioet, acmc-1cex9, ethanol, lithium salt 9ci, lithium 1+ ion ethoxide
PubChem CID 23661847
SMILES [Li+].CC[O-]

Specifications

Color Orange
Linear Formula LiOCH2CH3
Concentration Exact strength on the certificate of analysis
Density 0.8200g/mL
IUPAC Name lithium;ethanolate
Formula Weight 52
Identification Confirmed
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Lithium ethoxide, 1M solution in ethanol
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.