Learn More
Lithium hexafluorophosphate, 98%, pure, Thermo Scientific Chemicals
£32.66 - £354.77
Chemical Identifiers
CAS | 21324-40-3 |
---|---|
Molecular Formula | F6LiP |
Molecular Weight (g/mol) | 151.91 |
MDL Number | MFCD00011096 |
InChI Key | AXPLOJNSKRXQPA-UHFFFAOYSA-N |
Synonym | lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h |
PubChem CID | 23688915 |
IUPAC Name | lithium;hexafluorophosphate |
SMILES | [Li+].F[P-](F)(F)(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10589623
|
Thermo Scientific Acros
191260050 |
5 g | Plastic bottle |
£32.66
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10701091
|
Thermo Scientific Acros
191260250 |
25 g | Plastic bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388933
|
Thermo Scientific
191261000 |
100 g | Plastic bottle |
£354.77
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
21324-40-3 | |
151.91 | |
AXPLOJNSKRXQPA-UHFFFAOYSA-N | |
23688915 | |
[Li+].F[P-](F)(F)(F)(F)F |
F6LiP | |
MFCD00011096 | |
lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h | |
lithium;hexafluorophosphate |
Specifications
21324-40-3 | |
Fuming Powder | |
F6LiP | |
MFCD00011096 | |
Solubility in water: hydrolysis | |
[Li+].F[P-](F)(F)(F)(F)F | |
151.91 | |
151.91 | |
Pure |
White | |
97.5% min. (ex Li) | |
LiPF6 | |
lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h | |
AXPLOJNSKRXQPA-UHFFFAOYSA-N | |
lithium;hexafluorophosphate | |
23688915 | |
98% | |
Lithium hexafluorophosphate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-334-7
- Lithium hexafluorophosphate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.