missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluorophosphate, 98%

Product Code. 11327217
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 11327217

Brand: Thermo Scientific Alfa Aesar 011529.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used As An Electrolyte in Li-Ion Batteries | CAS: 21324-40-3 | F6LiP | 151.904 g/mol

Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21324-40-3
Molecular Formula F6LiP
Molecular Weight (g/mol) 151.904
MDL Number MFCD00011096
InChI Key AXPLOJNSKRXQPA-UHFFFAOYSA-N
Synonym lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h
PubChem CID 23688915
IUPAC Name lithium;hexafluorophosphate
SMILES [Li+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 200°C (decomposition)
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 98%
Solubility Information Soluble in water.
Formula Weight 151.9
Odor Odorless
Sensitivity Moisture sensitive
Appearance White powder
Density 1.5
Chemical Name or Material Lithium hexafluorophosphate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.