missing translation for 'onlineSavingsMsg'
Learn More
Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 12337123

Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 12337123
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 12337123

Brand: Thermo Scientific Acros 413321000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Boiling Point 920.0°C
Melting Point 462.0°C
Quantity 100 g
Assay Percent Range 98+%
Linear Formula LiOH·H2O
Merck Index 14,5534
Solubility Information Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol
IUPAC Name lithium(1+) hydrate hydroxide
Formula Weight 41.95
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Lithium hydroxide monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466763 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog Number413320025,413321000,413325000
Lot NumberA0456706 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number413320025,413321000,413325000
Lot NumberA0453153 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number413320025,413321000,413325000
Lot NumberA0455027 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number413320025,413321000,413325000
Lot NumberA0450824 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number413320025,413321000,413325000
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.