Learn More
Lithium iodide, 99%, pure, anhydrous, Thermo Scientific Chemicals
£166.00 - £575.00
Chemical Identifiers
CAS | 10377-51-2 |
---|---|
Molecular Formula | ILi |
Molecular Weight (g/mol) | 133.84 |
MDL Number | MFCD00011092 |
InChI Key | HSZCZNFXUDYRKD-UHFFFAOYSA-M |
Synonym | lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide |
PubChem CID | 66321 |
IUPAC Name | lithium;iodide |
SMILES | [Li+].[I-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10606422
|
Thermo Scientific Acros
203590500 |
50 g | Glass bottle |
£166.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10771861
|
Thermo Scientific Acros
203592500 |
250 g | Glass bottle |
£575.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10377-51-2 | |
133.84 | |
HSZCZNFXUDYRKD-UHFFFAOYSA-M | |
66321 | |
[Li+].[I-] |
ILi | |
MFCD00011092 | |
lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide | |
lithium;iodide |
Specifications
1171.0°C | |
10377-51-2 | |
98.5% min. (on anhydrous substance) (Argentometry) | |
LiI | |
01,615; 04,304; 05,410; 07,208; 09,283; 10,245; 11,300; 12,282; 15,185; 16,197; 17,85 | |
lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide | |
HSZCZNFXUDYRKD-UHFFFAOYSA-M | |
lithium;iodide | |
66321 | |
99% | |
3.49 |
446.0°C | |
Powder, Crystals and/or Granules | |
ILi | |
MFCD00011092 | |
15, 5591 | |
Solubility in water: 165 g/100 mL (20°C). Other solubilities: soluble in alcohol, freely soluble in acetone | |
[Li+].[I-] | |
133.84 | |
133.85 | |
Pure | |
Lithium iodide, 99% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 233-822-5
missing translation for 'tsca' : TSCA
- Lithium iodide, anhydrous
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : L-Limonene, 1, 8-Cineol, p-Cymene, D-Limonene
RUO – Research Use Only