Learn More
Lithium iodide hydrate, 99%, extra pure, Thermo Scientific Chemicals
£44.30 - £167.00
Chemical Identifiers
CAS | 85017-80-7 |
---|---|
Molecular Formula | ILi |
Molecular Weight (g/mol) | 133.84 |
MDL Number | MFCD00149769 |
InChI Key | HSZCZNFXUDYRKD-UHFFFAOYSA-M |
Synonym | lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci |
PubChem CID | 23688290 |
SMILES | [Li+].[I-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10441251
|
Thermo Scientific Acros
208011000 |
100 g | Glass bottle |
£44.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10286973
|
Thermo Scientific Acros
208015000 |
500 g | Glass bottle |
£167.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85017-80-7 | |
133.84 | |
HSZCZNFXUDYRKD-UHFFFAOYSA-M | |
23688290 |
ILi | |
MFCD00149769 | |
lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci | |
[Li+].[I-] |
Specifications
1171°C | |
85017-80-7 | |
Crystalline Powder and/or Crystals | |
ILi | |
MFCD00149769 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,freely soluble in acetone | |
[Li+].[I-] | |
133.84 | |
133.85 | |
Extra Pure |
73°C | |
White to Yellow | |
98.5% min. (on dry substance) (Argentometry) | |
LiI·xH2O | |
lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci | |
HSZCZNFXUDYRKD-UHFFFAOYSA-M | |
lithium;iodide;hydrate | |
23688290 | |
99% | |
Lithium iodide hydrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Warning
- Lithium iodide hydrate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only