missing translation for 'onlineSavingsMsg'
Learn More
Lithium L-lactate, 97% Product Code.: 11438980
  • ONLINE EXCLUSIVE PRICE
    £177.00
    £175.00 / 250g
    Save £2.00
    1% Off

Lithium L-lactate, 97%

Product Code. 11438980
250 g, 250g
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11438980

Brand: Thermo Scientific Alfa Aesar A11818.30

  • ONLINE EXCLUSIVE PRICE
    £177.00
    £175.00 / 250g
    Save £2.00
    1% Off
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27848-80-2 | C3H5LiO3 | 96.01 g/mol

Useful chiral synthon, building block for depsipeptides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27848-80-2
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065399
InChI Key GKQWYZBANWAFMQ-DKWTVANSSA-M
Synonym lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.liShow MoreShow Less
PubChem CID 23687877
IUPAC Name lithium;(2S)-2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Melting Point 253°C to 255°C
Quantity 250 g
Beilstein 3568332
Sensitivity Hygroscopic
Optical Rotation −14° (c=1 in water)
Formula Weight 96.01
Percent Purity 97%
Chemical Name or Material Lithium L-lactate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10252805 Certificate TypeCertificate of Analysis Date17/10/2024 Product CodeA11818.18,A11818.0B,A11818.30
Lot Number10247107 Certificate TypeCertificate of Analysis Date22/06/2023 Product CodeA11818.18,A11818.0B,A11818,A11818.30
Lot Number10237389 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeA11818.18,A11818.0B,A11818.30
Lot Number10239653 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeA11818.18,A11818.0B,A11818.30
Lot Number10216716 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11818.18,A11818.0B,A11818.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium L-lactate, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.