Learn More
Lithium molybdenum oxide, 99+%, Thermo Scientific Chemicals
£69.70 - £232.00
Chemical Identifiers
CAS | 13568-40-6 |
---|---|
Molecular Formula | Li2MoO4 |
Molecular Weight (g/mol) | 173.826 |
MDL Number | MFCD00016178 |
InChI Key | NMHMDUCCVHOJQI-UHFFFAOYSA-N |
Synonym | lithium molybdate, dilithium 1+ ion molybdate, dilithium dioxido dioxo molybdenum, dilithotab molybdate, 2li.moo4 |
PubChem CID | 6093689 |
IUPAC Name | dilithium;dioxido(dioxo)molybdenum |
SMILES | [Li+].[Li+].[O-][Mo](=O)(=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376758
|
Thermo Scientific Alfa Aesar
013427.22 |
100 g |
£69.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366758
|
Thermo Scientific Alfa Aesar
013427.36 |
500 g |
£232.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium molybdenum oxide is mainly used as an inhibitor in some types of industrial air conditioning. It is used in comparative property studies of battery systems. It has been used as a petroleum cracking catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13568-40-6 | |
173.826 | |
NMHMDUCCVHOJQI-UHFFFAOYSA-N | |
6093689 | |
[Li+].[Li+].[O-][Mo](=O)(=O)[O-] |
Li2MoO4 | |
MFCD00016178 | |
lithium molybdate, dilithium 1+ ion molybdate, dilithium dioxido dioxo molybdenum, dilithotab molybdate, 2li.moo4 | |
dilithium;dioxido(dioxo)molybdenum |
Specifications
705°C | |
White | |
Li2MoO4 | |
lithium molybdate, dilithium 1+ ion molybdate, dilithium dioxido dioxo molybdenum, dilithotab molybdate, 2li.moo4 | |
NMHMDUCCVHOJQI-UHFFFAOYSA-N | |
dilithium;dioxido(dioxo)molybdenum | |
6093689 | |
≥99% | |
Hygroscopic | |
Lithium molybdenum oxide |
13568-40-6 | |
Powder | |
MFCD00016178 | |
Soluble in water. | |
[Li+].[Li+].[O-][Mo](=O)(=O)[O-] | |
173.826 | |
173.83 | |
Odorless | |
2.66 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 236-977-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only