missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
Description
Lithium niobium ethoxide is used in the preparation of lithium niobate, which is a ferroelectric material used as an optical switch. It is also used as solid electrolytes due to its high ionic diffusivity in the field of material science.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Yellow |
Flash Point | 17°C (63°F) |
Odor | Alcohol-like |
Quantity | 25 mL |
UN Number | UN1170 |
Sensitivity | Moisture sensitive |
Formula Weight | 370.21 |
Percent Purity | ≥99% |
Physical Form | Liquid |
Assay | (metals basis) |
Show More |
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'