missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals

£62.60

Chemical Identifiers

CAS 86745-52-0
Molecular Formula C12H30LiNbO6
Molecular Weight (g/mol) 370.212
MDL Number MFCD00210625
InChI Key AVEKRHJBCCYCED-UHFFFAOYSA-N
Synonym lithium niobium ethoxide metals basis w/v in ethanol, lithium niobium ethoxide, lithium niobium 5+ ethoxide 1/1/6, niobium 5+ lithium 1+ hexakis ethoxide
PubChem CID 14693679
IUPAC Name lithium;ethanolate;niobium(5+)
SMILES [Li+].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323528
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039155.AC
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Lithium niobium ethoxide is used in the preparation of lithium niobate, which is a ferroelectric material used as an optical switch. It is also used as solid electrolytes due to its high ionic diffusivity in the field of material science.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86745-52-0
370.212
AVEKRHJBCCYCED-UHFFFAOYSA-N
14693679
[Li+].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
C12H30LiNbO6
MFCD00210625
lithium niobium ethoxide metals basis w/v in ethanol, lithium niobium ethoxide, lithium niobium 5+ ethoxide 1/1/6, niobium 5+ lithium 1+ hexakis ethoxide
lithium;ethanolate;niobium(5+)

Specifications

86745-52-0
17°C (63°F)
C12H30LiNbO6
UN1170
lithium niobium ethoxide metals basis w/v in ethanol, lithium niobium ethoxide, lithium niobium 5+ ethoxide 1/1/6, niobium 5+ lithium 1+ hexakis ethoxide
[Li+].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
370.212
370.21
Liquid
Lithium niobium ethoxide
Yellow
Alcohol-like
MFCD00210625
Moisture sensitive
AVEKRHJBCCYCED-UHFFFAOYSA-N
lithium;ethanolate;niobium(5+)
14693679
≥99%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

For Manufacturing and Laboratory Use. Not for beverage use