Learn More
Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol, Thermo Scientific Chemicals
£58.30 - £197.00
Chemical Identifiers
CAS | 21864-11-9 |
---|---|
MDL Number | MFCD00210627 |
Synonym | lithium niobium methoxide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313528
|
Thermo Scientific Alfa Aesar
039154.AC |
25 mL |
£58.30
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303528
|
Thermo Scientific Alfa Aesar
039154.AE |
100 mL |
£197.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as a precursor for chemical vapor deposition of thin films for lithium ion battery electrolyte materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
21864-11-9 | |
MFCD00210627 | |
lithium niobium methoxide |
Specifications
21864-11-9 | |
MFCD00210627 | |
Moisture sensitive | |
Fully miscible with water. | |
≥99% | |
(metals basis) |
LiNb(OCH3)6 | |
UN1230 | |
lithium niobium methoxide | |
285.92 | |
Liquid | |
Lithium niobium methoxide |
Safety and Handling
GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H311+H331-H314-H335-H370
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only