missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Nitrate Anhydrous, Extra Pure, SLR, Fisher Chemical™
103.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Specifications
Boiling Point | 600°C |
Melting Point | 251°C |
Color | White |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | lithium(1+) nitrate |
Formula Weight | 68.95g/mol |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS03_Oxidiser.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Lithium nitrate
- Warning
- Oxidising solid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'