Learn More
Lithium nitride, 99.4% (metals basis), Thermo Scientific Chemicals
£67.94 - £228.14
Chemical Identifiers
CAS | 26134-62-3 |
---|---|
Molecular Formula | Li3N |
Molecular Weight (g/mol) | 34.83 |
MDL Number | MFCD00016186 |
InChI Key | IDBFBDSKYCUNPW-UHFFFAOYSA-N |
Synonym | lithium nitride, trilithium nitride, li3n, lithium nitride li3n, nitrure de lithium, li3-n, nitruro de litio spanish, nitrure de lithium french, lithium nitride,anh, nli3 |
PubChem CID | 520242 |
SMILES | [Li]N([Li])[Li] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343637
|
Thermo Scientific Alfa Aesar
035479.09 |
10 g |
£67.94
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333637
|
Thermo Scientific Alfa Aesar
035479.18 |
50 g |
£228.14
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium nitride is used in metallurgy and chemical synthesis. It is also used to store hydrogen and acts as a source of the nitride ion. It is involved in the preparation of lithium hydride and lithium amide as well as a reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26134-62-3 | |
34.83 | |
IDBFBDSKYCUNPW-UHFFFAOYSA-N | |
520242 |
Li3N | |
MFCD00016186 | |
lithium nitride, trilithium nitride, li3n, lithium nitride li3n, nitrure de lithium, li3-n, nitruro de litio spanish, nitrure de lithium french, lithium nitride,anh, nli3 | |
[Li]N([Li])[Li] |
Specifications
845°C | |
Powder | |
Li3N | |
lithium nitride, trilithium nitride, li3n, lithium nitride li3n, nitrure de lithium, li3-n, nitruro de litio spanish, nitrure de lithium french, lithium nitride,anh, nli3 | |
IDBFBDSKYCUNPW-UHFFFAOYSA-N | |
Lithium nitride | |
520242 | |
Moisture sensitive | |
Lithium nitride |
26134-62-3 | |
99.4% (metals basis) | |
MFCD00016186 | |
Insoluble in organic solvents. | |
[Li]N([Li])[Li] | |
34.83 | |
34.82 | |
1.30 g/mL |
Safety and Handling
P210-P222-P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H250-H260-H314-H335
missing translation for 'dotInformation' : Hazard Class: 4.3; Packaging Group: I
missing translation for 'einecsNumber' : 247-475-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only