missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate, 95+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10547372
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 236.0°C
CAS 7791-03-9
Color White
pH 6.0 to 7.5 (5% soln. at 25°C)
Physical Form Crystalline Powder or Powder
Quantity 500 g
Assay Percent Range 95% min. (Argentometry)
Molecular Formula ClLiO4
Linear Formula LiClO4
MDL Number MFCD00011079
Fieser 17,167
Merck Index 15, 5595
Synonym lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
Solubility Information Solubility in water: 600g/L (25°C). Other solubilities: 1520g/L ethanol (25°C), 1820g/L methanol (25°C), 1140g/L ether (25°C), soluble in acetone and ethyl acetate
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES [Li+].[O-]Cl(=O)(=O)=O
IUPAC Name lithium;perchlorate
Molecular Weight (g/mol) 106.39
PubChem CID 23665649
Formula Weight 106.39
Percent Purity ≥95%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Lithium perchlorate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.