missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate, anhydrous, ACS, 95.0% min, Thermo Scientific Chemicals

Product Code. 11311748
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

As an oxidizing agent

Lithium perchlorate is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-03-9
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.43
MDL Number MFCD00011079
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
PubChem CID 23665649
IUPAC Name lithium;perchlorate
SMILES [Li+].[O-]Cl(=O)(=O)=O

Specifications

Boiling Point 400°C (decomposition)
Melting Point 236°C
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula LiClO4
UN Number UN1481
Merck Index 14,5539
Solubility Information Soluble in water,alcohol and ethyl acetate.
Formula Weight 106.4
Percent Purity ≤95.0%
Grade ACS
Sensitivity Hygroscopic
Density 2.43 g/mL
Chemical Name or Material Lithium perchlorate, Anhydrous
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.