missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium phthalocyanine

£50.83 - £237.93

Chemical Identifiers

CAS 25510-41-2
Molecular Formula C32H18Li2N8
Molecular Weight (g/mol) 528.432
MDL Number MFCD00059094
InChI Key VEGOPYLKEARSAH-UHFFFAOYSA-N
Synonym dilithium phthalocyanine
PubChem CID 66736323
SMILES [Li].[Li].C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378098
View Documents
Alfa Aesar™
039334.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11368098
View Documents
Alfa Aesar™
039334.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

25510-41-2
528.432
VEGOPYLKEARSAH-UHFFFAOYSA-N
66736323
C32H18Li2N8
MFCD00059094
dilithium phthalocyanine
[Li].[Li].C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3

Specifications

25510-41-2
C32H18Li2N8
dilithium phthalocyanine
[Li].[Li].C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3
66736323
Powder
>110°C (230°F)
MFCD00059094
VEGOPYLKEARSAH-UHFFFAOYSA-N
528.432
526.41
Lithium phthalocyanine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H360

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications