missing translation for 'onlineSavingsMsg'
Läs mer

Lithium salicylate, tech.

Produktkod. 11368118
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Förpackningsstorlek
100g
25g
500g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11368118

missing translation for 'mfr': Thermo Scientific Alfa Aesar 039356.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 552-38-5 | C7H5LiO3 | 144.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 552-38-5
Molecular Formula C7H5LiO3
Molecular Weight (g/mol) 144.054
MDL Number MFCD00045813
InChI Key PSBOOKLOXQFNPZ-UHFFFAOYSA-M
Synonym lithium salicylate, unii-93f1sp6qin, salicylic acid lithium salt, dilithium salicylate, 93f1sp6qin, lithium 2-hydroxybenzoate, lithium 1+ ion salicylate, lithiumsalicylate, lithiumsalicylat, salicylicacidlithiumsalt
PubChem CID 23663621
IUPAC Name lithium;2-hydroxybenzoate
SMILES [Li+].C1=CC=C(C(=C1)C(=O)[O-])O

Specifications

Melting Point (decomposition)
Odor Sweet
Quantity 500 g
Solubility Information Soluble in water
Formula Weight 144.05
Grade Technical
Physical Form Powder
Chemical Name or Material Lithium salicylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.