Learn More
Lithium sulfate, for analysis, anhydrous, Thermo Scientific Chemicals
£62.21 - £226.81
Chemical Identifiers
CAS | 10377-48-7 |
---|---|
Molecular Formula | Li2O4S |
Molecular Weight (g/mol) | 109.94 |
MDL Number | MFCD00011086 |
InChI Key | INHCSSUBVCNVSK-UHFFFAOYSA-L |
Synonym | lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrousShow More |
PubChem CID | 66320 |
ChEBI | CHEBI:53474 |
IUPAC Name | dilithium;sulfate |
SMILES | [Li+].[Li+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15344988
|
Thermo Scientific Acros
447552500 |
250 g |
£62.21
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15334988
|
Thermo Scientific Acros
447550010 |
1 kg |
£226.81
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10377-48-7 | |
109.94 | |
INHCSSUBVCNVSK-UHFFFAOYSA-L | |
66320 | |
dilithium;sulfate |
Li2O4S | |
MFCD00011086 | |
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrousShow More | |
CHEBI:53474 | |
[Li+].[Li+].[O-]S([O-])(=O)=O |
Specifications
859.0°C | |
Li2O4S | |
MFCD00011086 | |
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
INHCSSUBVCNVSK-UHFFFAOYSA-L | |
dilithium;sulfate | |
66320 | |
109.94 | |
2.22 | |
Lithium sulfate |
10377-48-7 | |
Li2SO4 | |
15,5597 | |
(10% in water) Clear colorless to slightly hazy | |
[Li+].[Li+].[O-]S([O-])(=O)=O | |
109.94 | |
CHEBI:53474 | |
Analytical | |
2.2200g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 233-820-4
- Lithium sulfate, anhydrous
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.