missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 845°C |
Color | White |
Physical Form | Coarse Powder |
Quantity | 100 g |
Merck Index | 14,5541 |
Solubility Information | Soluble in water(345g/L). Insoluble in absolute ethanol,acetone and pyridine. |
IUPAC Name | dilithium(1+) sulfate |
Formula Weight | 109.94 |
Percent Purity | 99.99% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'