missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Description
It act as a superconductor and widely used in lithium-sulfur batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 900°C to 975°C |
Color | Yellow |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.9% (Metal basis) |
Solubility Information | soluble in water and ethanol |
IUPAC Name | dilithium;sulfanide |
Formula Weight | 45.95 |
Odor | Like rotten eggs |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'