missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium tantalum ethoxide, 99.9% (metals basis), 5% w/v in ethanol, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Yellow |
Odor | Alcohol-like |
Linear Formula | LiTa(OCH2CH3)6 |
Quantity | 25 mL |
UN Number | UN1170 |
Sensitivity | Moisture Sensitive |
Formula Weight | 458.04 |
Percent Purity | 99.9% |
Physical Form | Liquid |
Assay | (metals basis) |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Lithium tantalum ethoxide 5% w/v in ethanol
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Ethyl alcohol, Lithium tantalum ethoxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'