missing translation for 'onlineSavingsMsg'
Learn More

Lithium tantalum isopropoxide, 99.9% (metals basis), Thermo Scientific™

Catalog Number 11348508
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
21864-24-4
MFCD00798538
This item is not returnable. View return policy
21864-24-4
MFCD00798538

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Boiling Point 160°C to 180°C (0.1 mmHg)
Linear Formula LiTa[OCH(CH3)2]6
Quantity 2 g
UN Number UN2925
Sensitivity Moisture sensitive
Formula Weight 542.42
Percent Purity 99.9%
Physical Form Wet Solid
Assay (metals basis)
Chemical Name or Material Lithium tantalum isopropoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'