missing translation for 'onlineSavingsMsg'
Learn More
Lithium tantalum isopropoxide, 99.9% (metals basis), Thermo Scientific™ Product Code.: 11368508

Lithium tantalum isopropoxide, 99.9% (metals basis), Thermo Scientific™

Product Code. 11368508
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11368508

Brand: Thermo Scientific Alfa Aesar 040520.09

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21864-24-4
MDL Number MFCD00798538

Specifications

Color White
Boiling Point 160°C to 180°C (0.1 mmHg)
Linear Formula LiTa[OCH(CH3)2]6
Quantity 10 g
UN Number UN2925
Sensitivity Moisture sensitive
Formula Weight 542.42
Percent Purity 99.9%
Physical Form Wet Solid
Assay (metals basis)
Chemical Name or Material Lithium tantalum isopropoxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium tantalum isopropoxide, 99.9% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.