Learn More
Lithium tert-butoxide, 99.9% (metals basis), Thermo Scientific Chemicals
£23.00 - £596.00
Chemical Identifiers
CAS | 1907-33-1 |
---|---|
Molecular Formula | C4H9LiO |
Molecular Weight (g/mol) | 80.055 |
MDL Number | MFCD00050479 |
InChI Key | LZWQNOHZMQIFBX-UHFFFAOYSA-N |
Synonym | lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate |
PubChem CID | 23664764 |
IUPAC Name | lithium;2-methylpropan-2-olate |
SMILES | [Li+].CC(C)(C)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389618
|
Thermo Scientific Alfa Aesar
044133.04 |
2 g |
£23.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379618
|
Thermo Scientific Alfa Aesar
044133.09 |
10 g |
£80.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369618
|
Thermo Scientific Alfa Aesar
044133.18 |
50 g |
£336.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399618
|
Thermo Scientific Alfa Aesar
044133.30 |
250 g | |||||||
Description
Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1907-33-1 | |
80.055 | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
23664764 | |
[Li+].CC(C)(C)[O-] |
C4H9LiO | |
MFCD00050479 | |
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate | |
lithium;2-methylpropan-2-olate |
Specifications
1907-33-1 | |
White to Cream | |
C4H9LiO | |
MFCD00050479 | |
3620018 | |
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
lithium;2-methylpropan-2-olate | |
23664764 | |
99.9% | |
(metals basis) |
∼150°C (decomposition) | |
Odorless | |
(CH3)3COLi | |
UN3206 | |
Air and moisture sensitive | |
Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether. | |
[Li+].CC(C)(C)[O-] | |
80.055 | |
80.06 | |
Powder | |
Lithium tert-butoxide |
Safety and Handling
GHS H Statement
H251-H314-H302
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.
P210-P240-P241-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H302-H314
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 217-611-5
missing translation for 'rtecsNumber' : UB8520000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only