Learn More
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
£112.00 - £1143.00
Chemical Identifiers
CAS | 1907-33-1 |
---|---|
Molecular Formula | C4H9LiO |
Molecular Weight (g/mol) | 80.06 |
InChI Key | LZWQNOHZMQIFBX-UHFFFAOYSA-N |
Synonym | lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate |
PubChem CID | 23664764 |
IUPAC Name | lithium;2-methylpropan-2-olate |
SMILES | [Li+].CC(C)(C)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10759633
|
Thermo Scientific Acros
301220250 |
25 g |
£112.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10609844
|
Thermo Scientific Acros
301221000 |
100 g |
£307.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397030
|
Thermo Scientific Acros
301225000 |
500 g |
£1143.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1907-33-1 | |
80.06 | |
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate | |
lithium;2-methylpropan-2-olate |
C4H9LiO | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
23664764 | |
[Li+].CC(C)(C)[O-] |
Specifications
1907-33-1 | |
0.8900g/mL | |
99% | |
C4H9LiO | |
2925 | |
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate | |
283 °C | |
[Li+].CC(C)(C)[O-] | |
80.06 | |
80.06 | |
Powder |
White to Yellow | |
Authentic | |
Plastic bottle | |
LiOCH(CH3)3 | |
0.89 | |
Solubility in water: vigorous reaction | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
lithium;2-methylpropan-2-olate | |
23664764 | |
99% | |
Lithium tert-butoxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Harmful if swallowed.
Reacts violently with water.
GHS P Statement
Keep cool.
Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-611-5
- Lithium tert-butoxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2-methyl-2-propanollithium salt
RUO – Research Use Only