missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium tetraborate trihydrate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed in the conductances of some potassium and sodium salts in dimethylformamide at 25°C.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Quantity | 500 g |
Merck Index | 14,5524 |
Solubility Information | Soluble in water. |
Formula Weight | 223.17 (169.12 Anhydrous) |
Percent Purity | 96% |
Chemical Name or Material | Lithium tetraborate trihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'