missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
2 g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Color | White |
Linear Formula | (C6H5)4BLi·3CH3OCH2CH2OCH3 |
Quantity | 0.5 g |
UN Number | UN1325 |
Sensitivity | Moisture sensitive |
Formula Weight | 595.54 (326.18 Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'