missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-tert-butoxyaluminohydride, 1.1M solution in THF, Thermo Scientific Chemicals

£92.16 - £516.57

Chemical Identifiers

CAS 17476-04-9, 109-99-9
Molecular Formula C12H28AlLiO3
MDL Number MFCD00011532
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10512833
View Documents
Thermo Scientific Acros
377581000
100 mL AcroSeal™ Glass Bottle in Tin Can
£92.16
100mL
Please to purchase this item. Need a web account? Register with us today!
10066132
View Documents Promotion Details
Thermo Scientific Acros
377588000
800 mL AcroSeal™ Glass Bottle
£516.57
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

17476-04-9, 109-99-9
C12H28AlLiO3
MFCD00011532

Specifications

Colorless
C12H28AlLiO3
0.9000g/mL
Solution
17476-04-9, 109-99-9
MFCD00011532
254.28
Lithium tri-tert-butoxyaluminohydride, 1.1M Solution in THF
Videos
Safety and Handling

Safety and Handling

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form exp

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present an

Danger

Recommended Storage : Product may develop turbidity during storage

Product Identifier
  • Lithium tri-tert-butoxyaluminohydride, 1.1M solution in THF

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only